Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atlastin-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26900625UL
Description
Atlastin-2 Polyclonal antibody specifically detects Atlastin-2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Atlastin-2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
ADP-ribosylation factor-like 6 interacting protein 2, ADP-ribosylation factor-like protein 6-interacting protein 2, ADP-ribosylation-like factor 6 interacting protein 2, aip-2, ARL3IP2, ARL-6-interacting protein 2, ARL6IP2, atlastin GTPase 2, atlastin2, atlastin-2, EC 3.6.5.-, FLJ23293 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RTSDPSAAVNHVSSTTSLGENYEDDDLVNSDEVMKKPCPVQIVLAHEDDHNFELDEEALEQ | |
25 μL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
64225 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction