Learn More
Atlastin-2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303333AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
ATL2 gene ontology annotations related to this gene include GTPase activity; GTP binding.Specifications
Atlastin-2 | |
Polyclonal | |
Unconjugated | |
ATL2 | |
GPHYSD1, ADAMTS-like protein 2, ADAMTSL-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64225 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8NHH9 | |
ATL2 | |
Between 533 and 583 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.