Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atlastin-3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297067APC
Description
Atlastin-3 Polyclonal antibody specifically detects Atlastin-3 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Atlastin-3 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
atlastin 3, atlastin GTPase 3, atlastin-3, DKFZP564J0863, EC 3.6.5.- | |
Rabbits immunized with purified, recombinant Human Atlastin-3 (Uniprot#: Q6DD88, Met1-Ala445) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
25923 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction