Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atlastin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215508
Description
Atlastin-3 Polyclonal specifically detects Atlastin-3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Atlastin-3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
atlastin 3, atlastin GTPase 3, atlastin-3, DKFZP564J0863, EC 3.6.5.- | |
Rabbit | |
61 kDa | |
0.1 mL | |
Primary | |
Expected to cross react based on sequence identity: Zebrafish (83%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q6DD88 | |
ATL3 | |
Recombinant protein encompassing a sequence within the center region of human ATL3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
25923 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction