Learn More
Atlastin-3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303313AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.Specifications
| Atlastin-3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q6DD88, Q91YH5 | |
| Atl3 | |
| Between 491 and 541 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ATL3 | |
| atlastin 3, atlastin-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109168, 25923 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.