Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM, p Ser1981 Antibody (10H11.E12), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 24 publications

$206.50 - $499.50

Specifications

Antigen ATM (p Ser1981)
Clone 10H11.E12
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100306SS
SDP
View Documents
Novus Biologicals
NB100306SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100306
SDP
View Documents
Novus Biologicals
NB100306
0.1 mL
Each for $499.50
Only null left
Add to cart
 
Description

Description

ATM Monoclonal specifically detects ATM in Human, Mouse, Rat, Canine (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

ATM (p Ser1981)
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse, Rat, Canine (Negative)
AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1
ATM
IgG1 κ
Protein G purified
Human, mouse and rat. Does not work in canine per customer review.
10H11.E12
Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Monoclonal
Purified
RUO
Q13315
472
Synthetic peptide made to a region surrounding the phosphorylated Serine 1981 of human ATM. [UniProt# Q13315]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
370 kDa
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.