Learn More
Invitrogen™ ATM Polyclonal Antibody, DyLight™ 550

Description
ATM Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, ELISA, IP
Specifications
Specifications
| Antigen | ATM |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | DyLight 550 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | ATM |
| Gene Accession No. | Q13315, Q62388 |
| Gene Alias | AI256621; AT mutated; A-T mutated; A-T mutated homolog; AT mutated protein; AT1; ATA; Ataxia t; Ataxia telangiectasia gene mutated in human beings; ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog; ATC; ATD; ATDC; ATE; Atm; ATM serine/threonine kinase; C030026E19Rik; DKFZp781A0353; EC 2.7.1.37; MGC74674; serine-protein kinase ATM; TEL1; TEL1, telomere maintenance 1, homolog; TELO1 |
| Gene Symbols | ATM |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.