Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATMIN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | ATMIN |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5026
![]() |
Bethyl Laboratories, Inc
A303399AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
The ATM/ATR-substrate CHK2-interacting zinc finger protein (ATMIN), also known as ASCIZ, forms DNA damage-induced nuclear foci that contain the DNA repair protein Rad51. ATMIN is also thought to be involved in embryonic development, as an absence of ATMIN causes late-embryonic lethality in mice with a range of organ development defects. It also activates the transcription DYNLL1, a light chain of the dynein motor complex and sequence-specific regulator of protein dimerization of numerous targets. DYNLL1 can bind to and inhibit the transcription activation domain of ATMIN, forming a simple dynamic feedback loop for DYNLL1 expression.Specifications
ATMIN | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
ASCIZ, ZNF822, KIAA0431, ATM interactor, Zinc finger protein 822, ASCIZ | |
ATMIN | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O43313 | |
23300 | |
Between 773 and 823 | |
Primary | |
4° C | |
ATMIN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title