Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATMIN Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5024 Supplier Bethyl Laboratories, Inc Supplier No. A303399A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The ATM/ATR-substrate CHK2-interacting zinc finger protein (ATMIN), also known as ASCIZ, forms DNA damage-induced nuclear foci that contain the DNA repair protein Rad51. ATMIN is also thought to be involved in embryonic development, as an absence of ATMIN causes late-embryonic lethality in mice with a range of organ development defects. It also activates the transcription DYNLL1, a light chain of the dynein motor complex and sequence-specific regulator of protein dimerization of numerous targets. DYNLL1 can bind to and inhibit the transcription activation domain of ATMIN, forming a simple dynamic feedback loop for DYNLL1 expression.

Specifications

Antigen ATMIN
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ATMIN
Gene Accession No. O43313
Gene Alias ASCIZ, ZNF822, KIAA0431, ATM interactor, Zinc finger protein 822, ASCIZ
Gene Symbols ATMIN
Host Species Rabbit
Immunogen Between 773 and 823
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23300
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.