Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATOH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144930
Description
Immunogen sequence: IYINALSELL QTPSGGEQPP PPPASCKSDH HHLRTAASYE GGAGNATAAG AQQASGGSQR PTPPGSCRTR FSAPASAGGY SVQLDALHFS TFEDSALTAM MAQKNLSPSL PGSILQPVQE ENSKTSPRSH RSDGEF Antigen sequence identity: Mouse 84%; Rat 83%.
The Drosophila atonal gene produces a protein with basic helix loop helix (bHLH) domains that plays an essential role in the development of the Drosophila nervous system. Mammalian atonal homolog 1 (MATH-1) is a helix-loop-helix (HLH) transcription factor that is structurally homologous to the product of the Drosophila proneural gene atonal. MATH-1, so known as Atoh1, Ath1 or HATH-1, is a 351 amino acid protein with an atonal-related basic HLH domain. In mice, expression of MATH-1 takes place by embryonic day 9. 5 and initially localizes to the cranial ganglions and the dorsal part of the central nervous system. Prominent expression of MATH-1 is in the dorsal part of the central nervous system but becomes restricted to the external granular layer of the cerebellum by day 18 and is undetectable in the adult nervous system. It is suggested that MATH-1 may play a role in the differentiation of subsets of neural cells by activating E box-dependent transcription.
Specifications
ATOH1 | |
Polyclonal | |
Unconjugated | |
Atoh1 | |
ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171 | |
Rabbit | |
Affinity chromatography | |
RUO | |
474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92858 | |
Atoh1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 206-341. Recombinant protein control fragment (Product #RP-110172). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction