Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATOH7 Monoclonal Antibody (2B6)
GREENER_CHOICE

Catalog No. PIMA523185
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523185 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523185 Supplier Invitrogen™ Supplier No. MA523185
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MQGLNTAFDR LRRVVPQWGQ DKKLSKYETL QMALSYIMAL TRILAEA.

ATOH7 is a member of the family of basic helix-loop-helix (bHLH) proteins with similarity to Drosophila atonal a proneural bHLH gene that controls photoreceptor development (Brown et al., 2002 [PubMed 11889557]). [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ATOH7
Gene Accession No. Q8N100
Gene Alias ath5; Atoh7; atonal bHLH transcription factor 7; atonal homolog 5; atonal homolog 7; atonal homolog 7 (Drosophila); atonal homolog bHLH transcription factor 7; BHLHA13; Class A basic helix-loop-helix protein 13; hATH5; Helix-loop-helix protein hATH-5; helix-loop-helix protein mATH-5; Helix-loop-helix protein zATH-5; lak; lakritz; Math5; NCRNA; PHPVAR; Protein atonal homolog 5; protein atonal homolog 7; Protein lakritz; RNANC; si:packtrz.1; zATH5
Gene Symbols ATOH7
Host Species Mouse
Immunogen ATOH7 (NP_660161, 53 a.a. ~ 99 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220202
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.