Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATOH7 Polyclonal Antibody
ATOH7 Polyclonal Antibody
Supplier: Thermo Scientific PA541212
Description
ATOH7 Polyclonal Antibody for Western Blot

Specifications
ATOH7 | |
Polyclonal | |
Unconjugated | |
ATOH7 | |
ath5; Atoh7; atonal bHLH transcription factor 7; atonal homolog 5; atonal homolog 7; atonal homolog 7 (Drosophila); atonal homolog bHLH transcription factor 7; BHLHA13; Class A basic helix-loop-helix protein 13; hATH5; Helix-loop-helix protein hATH-5; helix-loop-helix protein mATH-5; Helix-loop-helix protein zATH-5; lak; lakritz; Math5; NCRNA; PHPVAR; Protein atonal homolog 5; protein atonal homolog 7; Protein lakritz; RNANC; si:packtrz.1; zATH5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
220202 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N100 | |
ATOH7 | |
Synthetic peptide directed towards the C-terminal of human ATOH7 (aa 102-151). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction