Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATOH7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577183 Supplier Invitrogen™ Supplier No. PA577183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ATOH7 is a member of the family of basic helix-loop-helix (bHLH) proteins with similarity to Drosophila 'atonal,' a proneural bHLH gene that controls photoreceptor development (Brown et al., 2002 [PubMed 11889557]). [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ATOH7
Gene Accession No. Q8N100
Gene Alias ath5; Atoh7; atonal bHLH transcription factor 7; atonal homolog 5; atonal homolog 7; atonal homolog 7 (Drosophila); atonal homolog bHLH transcription factor 7; BHLHA13; Class A basic helix-loop-helix protein 13; hATH5; Helix-loop-helix protein hATH-5; helix-loop-helix protein mATH-5; Helix-loop-helix protein zATH-5; lak; lakritz; Math5; NCRNA; PHPVAR; Protein atonal homolog 5; protein atonal homolog 7; Protein lakritz; RNANC; si:packtrz.1; zATH5
Gene Symbols ATOH7
Host Species Rabbit
Immunogen Recombinant full length Human ATOH7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.