Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATOH7 Polyclonal Antibody

Catalog No. PIPA535859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535859 Supplier Invitrogen™ Supplier No. PA535859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: ATOH7-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ATOH7 is a member of the family of basic helix-loop-helix (bHLH) proteins with similarity to Drosophila 'atonal,' a proneural bHLH gene that controls photoreceptor development (Brown et al., 2002 [PubMed 11889557]). [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ATOH7
Gene Accession No. Q8N100, Q9Z2E5
Gene Alias ath5; Atoh7; atonal bHLH transcription factor 7; atonal homolog 5; atonal homolog 7; atonal homolog 7 (Drosophila); atonal homolog bHLH transcription factor 7; BHLHA13; Class A basic helix-loop-helix protein 13; hATH5; Helix-loop-helix protein hATH-5; helix-loop-helix protein mATH-5; Helix-loop-helix protein zATH-5; lak; lakritz; Math5; NCRNA; PHPVAR; Protein atonal homolog 5; protein atonal homolog 7; Protein lakritz; RNANC; si:packtrz.1; zATH5
Gene Symbols ATOH7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATOH7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220202, 53404
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.