Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATOH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565024 Supplier Invitrogen™ Supplier No. PA565024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKHIPVLEDGP WKTVCVKELN GLKKLKRKGK EPARRANGYK TFRLDLEAPE PRAVATNGLR DRTHRLQPVP VPVPVPVPYG APHR Highest antigen sequence identity to the following orthologs - mouse 73%, rat 72%.

Basic helix-loop-helix (bHLH) transcription factors play important roles in differentiation processes during embryonic development of vertebrates. ATOH8 (MATH6) is a tissue-restricted member of the atonal superfamily of bHLH transcription factors that exhibits 43-57% identity in the bHLH domain with other mammalian atonal paralogs including the NeuroD and Neurogenin factors. In the mouse, ATOH8 has been implicated in the specification and differentiation of neuronal cell lineages in the brain and may also participate in kidney development. Recent studies show that ATOH8 is a novel component of the pancreatic transcriptional network during embryonic development and suggest a potential role as a modulator of the differentiation program initiated by the pro-endocrine factor Neurog3. It is indispensable for early embryonic development, suggesting a more widespread function for this factor in tissue-specific differentiation processes that are dependent on class II bHLH genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATOH8
Gene Accession No. Q96SQ7
Gene Alias 4933425C05Rik; Ath6; Atoh8; atonal bHLH transcription factor 8; atonal homolog 6; atonal homolog 8; atonal homolog 8 (Drosophila); atonal homolog bHLH transcription factor 8; basic helix loop helix transcription factor 6; basic helix-loop-helix transcription factor 6; bHLHa21; Class A basic helix-loop-helix protein 21; hATH6; Helix-loop-helix protein hATH-6; helix-loop-helix protein mATH-6; mATH6; Okadin; Protein atonal homolog 8; RGD1561512
Gene Symbols ATOH8
Host Species Rabbit
Immunogen Recombinant Human ATOH8. Recombinant protein control fragment (Product #RP-105790).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.