Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATOX1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297082MFV450
Description
ATOX1 Polyclonal antibody specifically detects ATOX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
ATOX1 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
ATX1, ATX1 (antioxidant protein 1, yeast) homolog 1, ATX1 antioxidant protein 1 homolog (yeast), copper transport protein ATOX1, hah1, HAH1 MGC138453, Metal transport protein ATX1, MGC138455 | |
Rabbits immunized with purified, recombinant Human ATOX1 (Accession#: NP_004036.1, Met1-Glu68) | |
0.1 mL | |
Cancer, Cell Biology, Golgi Apparatus Markers, Signal Transduction | |
475 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction