Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATOX1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen ATOX1
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP106611SS
SDP
View Documents
Novus Biologicals
NBP106611SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP106611
SDP
View Documents
Novus Biologicals
NBP106611
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ATOX1 Polyclonal specifically detects ATOX1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ATOX1
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Cancer, Cell Biology, Golgi Apparatus Markers, Signal Transduction
Whole antisera with 0.1% Sodium Azide
ATX1, ATX1 (antioxidant protein 1, yeast) homolog 1, ATX1 antioxidant protein 1 homolog (yeast), copper transport protein ATOX1, hah1, HAH1 MGC138453, Metal transport protein ATX1, MGC138455
ATOX1
IgG
Unpurified
6 kDa
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Polyclonal
Antisera
RUO
Human
O00244
475
Full length human ATOX1 protein [Swiss-Prot# O00244]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.