Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL2/MCP1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $512.00
Specifications
Antigen | CCL2/MCP1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000. Use reported in scientific literature (PMID 22402584), Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 1:500-1:1000, Block/Neutralize reported in scientific literature (PMID 22778093) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP107035SS
![]() |
Novus Biologicals
NBP107035SS |
0.025 mL |
Each for $207.00
|
|
|||||
NBP107035
![]() |
Novus Biologicals
NBP107035 |
0.1 mL |
Each for $512.00
|
|
|||||
Description
CCL2/MCP1 Polyclonal specifically detects CCL2/MCP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
CCL2/MCP1 | |
Western Blot 1:1000. Use reported in scientific literature (PMID 22402584), Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 1:500-1:1000, Block/Neutralize reported in scientific literature (PMID 22778093) | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
6347 | |
Synthetic peptide made to an internal portion of rat MCP1 (within residues 15-40). [Swiss-Prot# P14844] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF | |
CCL2 | |
IgG | |
Affinity Purified | |
16 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title