Learn More
Invitrogen™ ATOX1 Monoclonal Antibody (4A12)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA518452
Catalog No. PIMA518452
Only null left
Add to Cart
Description
Description
Peptide Sequence: MPKHEFSVDM TCGGCAEAVS RVLNKLGGVK YDIDLPNKKV CIESEHSMDT LLATLKKTGK TVSYLGLE.
ATOX1 is a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in carcinogenesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATOX1
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
O00244
ATOX1
ATOX1 (NP_004036, 1 a.a. ~ 68 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
4A12
Unconjugated
ATOX1
antioxidant 1 copper chaperone; ATOX1; ATX1; ATX1 antioxidant protein 1 homolog; copper transport protein ATOX1; HAH1; Metal transport protein ATX1
Mouse
Protein A
RUO
475
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.