Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP Citrate Lyase Monoclonal Antibody (3A10)
GREENER_CHOICE

Catalog No. PIMA532947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532947 Supplier Invitrogen™ Supplier No. MA532947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP Citrate Lyase
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Acly
Gene Accession No. P53396
Gene Alias 17-beta-HSD 10; 17-beta-hydroxysteroid dehydrogenase 10; 17bHSD10; 17b-HSD10; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; A730098H14Rik; ABAD; AB-binding alcohol dehydrogenase; ACL; ACLY; Ads9; amyloid beta-peptide binding protein; amyloid-beta peptide binding alcohol dehydrogenase; ATP citrate lyase; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; ATPCL; AW538652; CAMR; citrate cleavage enzyme; CLATP; DUPXp11.22; endoplasmic reticulum-associated amyloid beta-peptide-binding protein; ERAB; HADH2; HCD2; Hsd17b10; hydroxyacyl-Coenzyme A dehydrogenase type II; hydroxyacyl-Coenzyme A dehydrogenase, type II; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid 17-beta dehydrogenase 10; hydroxysteroid dehydrogenase 10; MHBD; Mitochondrial ribonuclease P protein 2; mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; MRPP2; MRX17; MRX31; MRXS10; SCHAD; SDR5C1; short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; Type II HADH; XH98G2
Gene Symbols Acly
Host Species Mouse
Immunogen Synthetic peptide, corresponding to amino acids C-terminus of Human ATP-Citrate Lyase.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 47
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.