Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP Citrate Lyase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578712
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578712 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578712 Supplier Invitrogen™ Supplier No. PA578712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human MOLT4 whole cell, human U87 whole cell, human U251 whole cell, human 293T whole cell, human Hela whole cell, human T47D whole cell, human HepG2 whole cell, rat pancreas tissue, mouse NIH/3T3 whole cell. IHC: human colonic adenocarcinoma tissue, human hepatocellular carcinoma tissue, human laryngeal squamous cell carcinoma tissue, human placenta tissue, human colon cancer tissue, mouse testis tissue, rat testis tissue. ICC/IF: A594 cell. Flow: HepG2 cell.

ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP Citrate Lyase
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Acly
Gene Accession No. P16638, P53396, Q91V92
Gene Alias 17-beta-HSD 10; 17-beta-hydroxysteroid dehydrogenase 10; 17bHSD10; 17b-HSD10; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; A730098H14Rik; ABAD; AB-binding alcohol dehydrogenase; ACL; ACLY; Ads9; amyloid beta-peptide binding protein; amyloid-beta peptide binding alcohol dehydrogenase; ATP citrate lyase; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; ATPCL; AW538652; CAMR; citrate cleavage enzyme; CLATP; DUPXp11.22; endoplasmic reticulum-associated amyloid beta-peptide-binding protein; ERAB; HADH2; HCD2; Hsd17b10; hydroxyacyl-Coenzyme A dehydrogenase type II; hydroxyacyl-Coenzyme A dehydrogenase, type II; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid 17-beta dehydrogenase 10; hydroxysteroid dehydrogenase 10; MHBD; Mitochondrial ribonuclease P protein 2; mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; MRPP2; MRX17; MRX31; MRXS10; SCHAD; SDR5C1; short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; Type II HADH; XH98G2
Gene Symbols Acly
Host Species Rabbit
Immunogen E. coli-derived human ATP citrate lyase recombinant protein (Position: M1-I180).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104112, 24159, 47
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.