Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP5G1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568102
Description
Predicted to react with Mouse and Rat samples.
Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. F1 consists of five distinct subunits, designated ATP5A, ATP5B, ATP5C1, ATP5D and ATP5E, while F0 consists of ten subunits, designated ATP5H, ATP5G1, ATP5I, ATP5G2, ATP5J2, ATP5J, ATP5G3, ATP5S, ATP5F1 and ATP5L. ATP5G1, ATP5G2 and ATP5G3 localize to the mitochondrial membrane and exist as subunits of the F0 complex. ATP5G2 exists as three alternatively spliced isoforms.
Specifications
ATP5G1 | |
Polyclonal | |
Unconjugated | |
ATP5MC1 | |
ATP Synthase C; ATP synthase F(0) complex subunit C1, mitochondrial; ATP synthase H+ transporting mitochondrial F0 complex subunit c (subunit 9) isoform 1; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 1; ATP synthase proteolipid P1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9); ATP5A; ATP5G; ATP5G1; ATP5MC1; ATPase protein 9; ATPase subunit 9; ATPase subunit c; mitochondrial ATP synthase, subunit 9, isoform 1; mitochondrial ATP synthase, subunit C, isoform 1 | |
Rabbit | |
affinity chromatography | |
RUO | |
11951, 29754, 516 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05496, Q06645, Q9CR84 | |
ATP5MC1 | |
Synthesized peptide derived from human ATP Synthase C. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction