Learn More
Invitrogen™ ATP Synthase gamma Monoclonal Antibody (OTI2H6)

Description
ATP Synthase gamma Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ATP Synthase gamma |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2H6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ATP5F1C |
| Gene Accession No. | P36542 |
| Gene Alias | 1700094F02Rik; ATP synthase F1 subunit gamma; ATP synthase gamma chain, mitochondrial; ATP synthase gamma-subunit gene encoding a mitochondrial protein; ATP synthase subunit gamma, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1; ATP5C; Atp5c1; ATP5CL1; ATP5F1C; F1 gamma; F-ATPase gamma subunit; mitochondrial ATP synthase, gamma subunit 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.