Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP Synthase O Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559548 Supplier Invitrogen™ Supplier No. PA559548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEEATLSELK TVLKSFLSQG QVLKLEAKTD PSILGGMIVR IGEKYVDMSV KTKIQKLGRA MREIV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP Synthase O
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP5PO
Gene Accession No. P48047
Gene Alias ATP synthase peripheral stalk subunit OSCP; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein); ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; ATP5O; ATP5PO; ATPO; D12Wsu28e; HMC08D05; human ATP synthase OSCP subunit; oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; OSCP; Sperm flagella protein 4
Gene Symbols ATP5PO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP Synthase O. Recombinant protein control fragment (Product #RP-95885).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.