Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP Synthase O Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568104
Description
Predicted to react with Mouse and Rat samples.
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
Specifications
ATP Synthase O | |
Polyclonal | |
Unconjugated | |
ATP5PO | |
ATP synthase peripheral stalk subunit OSCP; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein); ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; ATP5O; ATP5PO; ATPO; D12Wsu28e; HMC08D05; human ATP synthase OSCP subunit; oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; OSCP; Sperm flagella protein 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
192241, 28080, 539 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48047, Q06647, Q9DB20 | |
ATP5PO | |
Synthesized peptide derived from human ATP Synthase O. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction