Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120400 Supplier Invitrogen™ Supplier No. PA5120400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, U-251MG, DU-145, OVCAR3, HT-29, A-431. The target is usually found in the following locations: Cell membrane, Endoplasmic reticulum membrane, Multi-pass membrane protein. Immunogen sequence: RRCTVSGVEY SHDANAQRLA RYQEADSEEE EVVPRGGSVS QRGSIGSHQS VRVVHRTQST KSHRRTGSRA EAKRASMLSK HTAFSSPMEK DITPDPKLLE KVSECDKSLA VARHQEHLLA HLSPELSDVF DFFIALTICN TVVVTSPDQP RTKVRVRFEL KSPVKTIEDF LRRFTPSCLT SGCSSIGSLA ANKSSHKLGS SFPSTPSSDG MLLRLEERLG QPTSAIASNG YSSQADNWAS ELAQEQESER E.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP10A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ATP10A
Gene Accession No. O60312
Gene Alias aminophospholipid translocase VA; ATP10A; ATP10C; ATPase class V type 10A; ATPase phospholipid transporting 10A (putative); ATPase type IV, phospholipid transporting (P-type); ATPase, class V, type 10A; ATPase, Class V, type 10C; ATPVA; ATPVC; KIAA0566; P4-ATPase flippase complex alpha subunit ATP10A; phospholipid-transporting ATPase VA; probable phospholipid-transporting ATPase VA
Gene Symbols ATP10A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 440-690 of human ATP10A (NP_077816.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57194
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.