Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559593 Supplier Invitrogen™ Supplier No. PA559593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTQLQLARQL TRKSPRRCSA PKETFAQGRL PKDSGTEHSS GRTVKTSVPL SQPSWHTQQP VCSLEASGEP STVDMSMPVR EHTLLE Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP10A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP10A
Gene Accession No. O60312
Gene Alias aminophospholipid translocase VA; ATP10A; ATP10C; ATPase class V type 10A; ATPase phospholipid transporting 10A (putative); ATPase type IV, phospholipid transporting (P-type); ATPase, class V, type 10A; ATPase, Class V, type 10C; ATPVA; ATPVC; KIAA0566; P4-ATPase flippase complex alpha subunit ATP10A; phospholipid-transporting ATPase VA; probable phospholipid-transporting ATPase VA
Gene Symbols ATP10A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP10A. Recombinant protein control fragment (Product #RP-98195).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57194
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.