Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP10D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569611 Supplier Invitrogen™ Supplier No. PA569611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 91%.

Catalytic component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP10D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATP10D
Gene Accession No. 0
Gene Alias 9830145H18Rik; Atp10d; ATPase class V type 10D; ATPase phospholipid transporting 10D; ATPase phospholipid transporting 10D (putative); ATPase, class V, type 10D; ATPVD; D5Buc24e; KIAA1487; P4-ATPase flippase complex alpha subunit ATP10D; probable phospholipid-transporting ATPase VD; putative type IV aminophospholipid transporting ATPase; si:dkey-20i10.6; type IV aminophospholipid transporting ATPase
Gene Symbols ATP10D
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat ATP10D.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360932
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.