Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP10D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562245 Supplier Invitrogen™ Supplier No. PA562245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFPSPILRAK HFDRLTPEER TKALKKWRGA GKMNQVTSKY ANQSAGKSGR RPMPGPSAVF AMKSASSCAI EQGNLSLCET ALDQGYSETK AFEMAGPS Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 63%.

Catalytic component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP10D
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP10D
Gene Accession No. Q9P241
Gene Alias 9830145H18Rik; Atp10d; ATPase class V type 10D; ATPase phospholipid transporting 10D; ATPase phospholipid transporting 10D (putative); ATPase, class V, type 10D; ATPVD; D5Buc24e; KIAA1487; P4-ATPase flippase complex alpha subunit ATP10D; probable phospholipid-transporting ATPase VD; putative type IV aminophospholipid transporting ATPase; si:dkey-20i10.6; type IV aminophospholipid transporting ATPase
Gene Symbols ATP10D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP10D. Recombinant protein control fragment (Product #RP-96674).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.