Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557324 Supplier Invitrogen™ Supplier No. PA557324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCYACKLFRR NTQLLELTTK RIEEQSLHDV LFELSKTVLR HSGSLTRDNL SGLSADMQDY GLIIDGAALS LIMKPREDGS SGNYRELFLE ICRSC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

ATP11A is a widely expressed integral membrane ATPase. ATP11A is probably phosphorylated in its intermediate state and likely drives the transport of ions such as calcium or other molecules across membranes. While the exact molecule ATP11A transports is unknown, increased expression of ATP11A mRNA has been observed in murine leukemia cells made resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs). Furthermore, overexpression of ATP11A provided protection against the FTI SCH66336 while knockdown of ATP11A via siRNA made cells more sensitive to this drug. Other reports suggest that elevated levels of ATP11A mRNA may also be a predictive marker of metastasis in colorectal cancer patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP11A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP11A
Gene Accession No. P98196
Gene Alias 4930558F19Rik; ATP11A; ATPase 11A, class VI; ATPase 11A, p type; ATPase class VI type 11A; ATPase IS; ATPase phospholipid transporting 11A; ATPase, class 1, member h; ATPase, class VI, type 11A; Atpc1h; ATPIH; ATPIS; AU040868; Ih; KIAA1021; P4-ATPase flippase complex alpha subunit ATP11A; phospholipid-translocating ATPase; potential phospholipid-transporting ATPase IH; probable phospholipid-transporting ATPase IH; RP11-120K24.1; Ua20
Gene Symbols ATP11A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP11A. Recombinant protein control fragment (Product #RP-94972).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.