Learn More
Invitrogen™ ATP11B Monoclonal Antibody (4H8)

Description
Peptide Sequence: DIIKKVFDRH LHPTSTEKAQ LTETNAGIKC LDSMCCFPEG EAACASVGRM LERVIGRCSP THISRSWSAS DPFYTNDRSI LTLSTMDSST C.
Specifications
Specifications
| Antigen | ATP11B |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4H8 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ATP11B |
| Gene Accession No. | Q9Y2G3 |
| Gene Alias | 1110019I14Rik; ATP11B; ATPase class VI type 11B; ATPase IR; ATPase phospholipid transporting 11B (putative); ATPase, class VI, type 11B; ATPIF; ATPIR; KIAA0956; mKIAA0956; P4-ATPase flippase complex alpha subunit ATP11B; Probable phospholipid-transporting ATPase IF; TIEG; TIEG1; truncated ATPase 11B protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.