Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP11B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520996

Catalog No. PIPA520996


Only null left
Add to Cart

Description

Description

A suggested positive control is K562 cell lysate. PA5-20996 can be used with blocking peptide PEP-1110.

ATP11B is a widely expressed integral membrane ATPase and is thought to drive the transport of phospholipids across membranes. It has been suggested that ATP11B is hormonally regulated and plays a role in the subnuclear trafficking of transcription factors with RING motifs. While the exact molecule ATP11B transports is unknown, increased mRNA expression of the homologous ATP11A has been observed in cells resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs), suggesting that ATP11B may also play a role in cell survival under harsh conditions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP11B
Polyclonal
Unconjugated
ATP11B
1110019I14Rik; ATP11B; ATPase class VI type 11B; ATPase IR; ATPase phospholipid transporting 11B (putative); ATPase, class VI, type 11B; ATPIF; ATPIR; KIAA0956; mKIAA0956; P4-ATPase flippase complex alpha subunit ATP11B; Probable phospholipid-transporting ATPase IF; TIEG; TIEG1; truncated ATPase 11B protein
Rabbit
Antigen Affinity Chromatography
RUO
23200, 76295
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q9Y2G3
ATP11B
A 19 amino acid peptide near the amino terminus of human ATP11B.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.