Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP11C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556569 Supplier Invitrogen™ Supplier No. PA556569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQHINIDTLT SDPRLYMKIS GNAMLQLG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 89%.

The ATP11C gene encodes a protein that belongs to the P4-ATPase family, specifically functioning as a phospholipid-transporting ATPase. ATP11C plays a critical role in the maintenance of phospholipid asymmetry across the cell membrane by transporting aminophospholipids from the outer leaflet to the inner leaflet of the plasma membrane. This activity is essential for various biological processes, including cell membrane integrity, cell signaling, and apoptosis. ATP11C is particularly important in hematopoiesis, influencing the development of B cells and red blood cells. Mutations or dysregulation in ATP11C can lead to disruptions in cell membrane dynamics and are associated with immunological disorders and anemia. Research on ATP11C continues to focus on its role in phospholipid transport and its implications for cellular function and disease, particularly in the context of blood-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP11C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Atp11c
Gene Accession No. Q8NB49
Gene Alias A330005H02Rik; AI315324; Atp11c; Atpase 11c, p type; ATPase class VI type 11C; ATPase IQ; ATPase phospholipid transporting 11C; ATPase, class VI, type 11C; ATPIG; ATPIQ; I79_007666; Ig; P4-ATPase flippase complex alpha subunit ATP11C; phospholipid-transporting ATPase 11C; Phospholipid-transporting ATPase IG; probable phospholipid-transporting ATPase 11C; probable phospholipid-transporting ATPase IG; putative phospholipid-transporting ATPase 11C
Gene Symbols Atp11c
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP11C. Recombinant protein control fragment (Product #RP-95435).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286410
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.