Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP11C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525361
Description
ATP11C Polyclonal Antibody for Western Blot, IHC (P), Flow
The ATP11C gene encodes a protein that belongs to the P4-ATPase family, specifically functioning as a phospholipid-transporting ATPase. ATP11C plays a critical role in the maintenance of phospholipid asymmetry across the cell membrane by transporting aminophospholipids from the outer leaflet to the inner leaflet of the plasma membrane. This activity is essential for various biological processes, including cell membrane integrity, cell signaling, and apoptosis. ATP11C is particularly important in hematopoiesis, influencing the development of B cells and red blood cells. Mutations or dysregulation in ATP11C can lead to disruptions in cell membrane dynamics and are associated with immunological disorders and anemia. Research on ATP11C continues to focus on its role in phospholipid transport and its implications for cellular function and disease, particularly in the context of blood-related disorders.
Specifications
ATP11C | |
Polyclonal | |
Unconjugated | |
Atp11c | |
A330005H02Rik; AI315324; Atp11c; Atpase 11c, p type; ATPase class VI type 11C; ATPase IQ; ATPase phospholipid transporting 11C; ATPase, class VI, type 11C; ATPIG; ATPIQ; I79_007666; Ig; P4-ATPase flippase complex alpha subunit ATP11C; phospholipid-transporting ATPase 11C; Phospholipid-transporting ATPase IG; probable phospholipid-transporting ATPase 11C; probable phospholipid-transporting ATPase IG; putative phospholipid-transporting ATPase 11C | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100756336, 286410, 320940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NB49, Q9QZW0 | |
Atp11c | |
KLH conjugated synthetic peptide between 589-616 amino acids from the central region of human ATP11C. | |
400 μL | |
Primary | |
Human, Mouse, Hamster | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction