Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549624
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549624 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549624 Supplier Invitrogen™ Supplier No. PA549624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP12A Polyclonal Antibody for Western Blot, IHC (P), Flow

Catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. Responsible for potassium absorption in various tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP12A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ATP12A
Gene Accession No. P54707
Gene Alias #945; ATP12A; ATP1AL1; ATPase H+/K+ transporting non-gastric alpha2 subunit; ATPase H+K+-transporting, alpha 2; ATPase, H+/K+ transporting, nongastric, alpha polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 1; cHKA; HK; HKalpha2; non-gastric H(+)/K(+) ATPase alpha subunit; non-gastric H(+)/K(+) ATPase subunit alpha; Potassium-transporting ATPase alpha chain 2; proton pump
Gene Symbols ATP12A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 833-859 amino acids from the Central region of human ATP12A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.