Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP13A2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11041486C
Description
ATP13A2 Polyclonal specifically detects ATP13A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ATP13A2 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
Q9NQ11 | |
ATP13A2 | |
A synthetic peptide made to an internal region of the human ATP13A2 protein (within residues 200-300). [Swiss-Prot# Q9NQ11] | |
0.1 mL | |
Cancer, Neurodegeneration, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
23400 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATPase type 13A2, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.5, EC 3.6.3.8, FLJ26510, HSA9947, KRPPD, PARK9putative ATPase, Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome), probable cation-transporting ATPase 13A2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction