Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP13A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
$589.50
Specifications
| Antigen | ATP13A2 |
|---|---|
| Concentration | 0.40 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111322
![]() |
Invitrogen™
PA5111322 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111398
![]() |
Invitrogen™
PA5111398 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: EIRDKEDSSW QLFTVQVQTE AIGEGSLEPS PQSQAEDGRS QAAVGAVPEG AWKDTAQLHK SEEAVSVGQK RVLRYYLFQG QRYIWIET.
Neurodegenerative disorders such as Parkinson and Alzheimer disease cause motor and cognitive dysfunction and belong to a heterogeneous group of common and disabling disorders. ATP13A2, otherwise known as PARK9, is a neuronal P-type ATPase gene underlying an autosomal recessive form of early-onset parkinsonism with pyramidal degeneration and dementia. ATP13A2 protein is located in the membrane of these lysosomes and is formed most strongly in the brain, especially in the substantia nigra, a brain region which is known to play a central role in Parkinson's disease.Specifications
| ATP13A2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ATP13A2 | |
| 1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4 | |
| ATP13A2 | |
| Primary | |
| Antigen Affinity Chromatography |
| 0.40 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ11 | |
| 23400 | |
| Recombinant Protein corresonding to Human ATP13A2. Recombinant protein control fragment (Product #RP-103955). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title