Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP13A2 Polyclonal Antibody

Catalog No. PIPA5111322
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111322 100 μL
PIPA5111398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111322 Supplier Invitrogen™ Supplier No. PA5111322

Rabbit Polyclonal Antibody

Immunogen sequence: EIRDKEDSSW QLFTVQVQTE AIGEGSLEPS PQSQAEDGRS QAAVGAVPEG AWKDTAQLHK SEEAVSVGQK RVLRYYLFQG QRYIWIET Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 69%.

Neurodegenerative disorders such as Parkinson and Alzheimer disease cause motor and cognitive dysfunction and belong to a heterogeneous group of common and disabling disorders. ATP13A2, otherwise known as PARK9, is a neuronal P-type ATPase gene underlying an autosomal recessive form of early-onset parkinsonism with pyramidal degeneration and dementia. ATP13A2 protein is located in the membrane of these lysosomes and is formed most strongly in the brain, especially in the substantia nigra, a brain region which is known to play a central role in Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP13A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP13A2
Gene Accession No. Q9NQ11
Gene Alias 1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4
Gene Symbols ATP13A2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ATP13A2. Recombinant protein control fragment (Product #RP-103955).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.