Learn More
Invitrogen™ ATP13A2 Polyclonal Antibody

Description
Immunogen sequence: EIRDKEDSSW QLFTVQVQTE AIGEGSLEPS PQSQAEDGRS QAAVGAVPEG AWKDTAQLHK SEEAVSVGQK RVLRYYLFQG QRYIWIET Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 69%.
Specifications
Specifications
| Antigen | ATP13A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ATP13A2 |
| Gene Accession No. | Q9NQ11 |
| Gene Alias | 1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4 |
| Gene Symbols | ATP13A2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.