Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587469 Supplier Invitrogen™ Supplier No. PA587469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVLGFCHLFL PDEQFPEGFQ FDTDDVNFPI DNLCFVGLIS MIDPPRAAVP DAVGKCRSAG IKVIMVTGDH PITAKAIAKG VGIISEGNET VEDIAARLNI PVSQVNPRDA KACVVHGSDL KDMTSEQLDD ILKYHTEIVF ARTSPQQKLI IVEGCQRQGA IVAVTGDGVN DSPALKKADI GVAMGIAGSD VSKQAADMIL LDDNFASIVT GVEEGRLIFD NLKKSIAYTL TSNIPEITPF LIFIIANIPL PLGTVTILCI DLGTDMVPAI SLAYEQAESD IMKRQPRNPK TDKLVNERLI; Positive Samples: LO2; Cellular Location: Cell membrane, Melanosome, Multi-pass membrane protein.

The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 1 subunit. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1A1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP1A1
Gene Accession No. P05023
Gene Alias (Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; alpha subunit of sodium potassium ATPase; atp1a1; atp1a1-a; atp1a1a.4; atp1a1-b; ATP1B; Atpa-1; ATPase; ATPase Na+/K+ transporting subunit alpha 1; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; BC010319; K-ATPase; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na, K-ATPase alpha-1 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 isoform; Na/K ATPase alpha 1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na+K+ATPase; Nkaa1b; RP4-655J12.1; sarcolemmal sodium/potassium-ATPase alpha 1; Sodium Potassium ATPase; sodium pump 1; sodium pump subunit alpha-1; sodium/potassium-ATPase alpha-subunit 1; sodium/potassium-transporting ATPase alpha-1 chain; sodium/potassium-transporting ATPase subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; unnamed protein product
Gene Symbols ATP1A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 551-850 of human ATP1A (NP_0006922).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 476
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.