Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1A2 Recombinant Rabbit Monoclonal Antibody (5T3O4)
GREENER_CHOICE

Catalog No. PIMA537905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537905 Supplier Invitrogen™ Supplier No. MA537905
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse heart, Mouse skeletal muscle, Rat heart Immunogen sequence: MGRGAGREYS PAATTAENGG GKKKQKEKEL DELKKEVAMD DHKLSLDELG RKYQVDLSKG LTNQRAQDVL ARDGPNALTP PPTTPEWVKF CRQLFGGFSI.

Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1A2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5T3O4
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Atp1a2
Gene Accession No. P06686, Q6PIE5
Gene Alias Atp1a2; Atpa-3; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase Na+/K+ transporting subunit alpha 2; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; AW060654; FHM2; KIAA0778; MHP2; mKIAA0778; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; RATATPA2; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; sodium/potassium-transporting ATPase subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2
Gene Symbols Atp1a2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ATP1A2 (P50993).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24212, 98660
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.