Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572353
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572353 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572353 Supplier Invitrogen™ Supplier No. PA572353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP1A4 Polyclonal Antibody for Western Blot, Flow

This is the catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. This action creates the electrochemical gradient of sodium and potassium ions, providing the energy for active transport of various nutrients. Plays a role in sperm motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1A4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ATP1A4
Gene Accession No. Q13733
Gene Alias 3.6.3.9; ATP1A1; ATP1A4; ATP1AL2; ATPase Na+/K+ transporting subunit alpha 4; ATPase, Na+/K+ transporting, alpha 4 polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 2; Na(+)/K(+) ATPase alpha-4 subunit; Na,K-ATPase subunit alpha-C; Na+/K+ ATPase 4; Na+/K+ ATPase alpha 4 subunit; Na+/K+ ATPase, alpha-D polypeptide; Sodium Potassium ATPase; sodium pump 4; sodium pump subunit alpha-4; sodium/potassium-transporting ATPase alpha-4 chain; sodium/potassium-transporting ATPase subunit alpha-4; sodium-potassium ATPase catalytic subunit alpha-4
Gene Symbols ATP1A4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 12-46 amino acids from the human region of human ATP1A4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.