Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1B1 Monoclonal Antibody (464.8 (aka 8A))
GREENER_CHOICE

Catalog No. AMPA6732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6732 Supplier Invitrogen™ Supplier No. MA116732
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with mouse samples.

The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 1 subunit. Alternatively spliced transcript variants encoding different isoforms have been described, but their biological validity is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 464.8 (aka 8A)
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.02% sodium azide
Gene ATP1B1
Gene Accession No. P05026, P05027, P06583, P07340, P08251, P14094, Q9TT37
Gene Alias adenosinetriphosphatase; ATP1B; ATP1B1; Atp4b; ATPase beta subunit; ATPase Na+/K+ transporting subunit beta 1; ATPase, Na+/K+ transporting, beta 1 polypeptide; Atpb; Atpb-1; ATPBS; Beta 1-subunit of Na(+),K(+)-ATPase; hypothetical protein LOC519758; MGC1798; Na, K-ATPase beta-1 polypeptide; Na,K-ATPase beta 1 subunit; Na+, K+-ATPase; Na+, K+-ATPase beta-subunit; Na+,K+-ATPase beta 1 subunit; Na+/K+ ATPase Beta; NKbeta1; OTTHUMP00000032537; OTTHUMP00000032538; P-type ATPase; Sodium Potassium ATPase; sodium pump subunit beta-1; sodium/potassium ATPase beta subunit; sodium/potassium-dependent ATPase beta-1 subunit; sodium/potassium-dependent ATPase subunit beta-1; sodium/potassium-transporting ATPase beta-1 chain; sodium/potassium-transporting ATPase subunit beta-1; sodium-potassium ATPase subunit beta 1 (non-catalytic); unnamed protein product
Gene Symbols ATP1B1
Host Species Mouse
Immunogen Purified Sodium Potassium ATPase Beta 1 from rabbit renal outer medulla.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008749, 100760718, 11931, 25650, 396529, 396898, 403966, 481, 519758
Target Species Bovine, Canine, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Frog
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.