Learn More
Invitrogen™ ATP1B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529719
Description
Recommended positive controls: K562, HL-60, mouse brain, Rat brain. Predicted reactivity: Mouse (90%), Rat (92%), Dog (91%), Pig (89%), Rabbit (87%), Sheep (91%), Rhesus Monkey (96%), Chimpanzee (99%), Guinea pig (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 1 subunit. Alternatively spliced transcript variants encoding different isoforms have been described, but their biological validity is not known.
Specifications
| ATP1B1 | |
| Polyclonal | |
| Unconjugated | |
| ATP1B1 | |
| adenosinetriphosphatase; ATP1B; ATP1B1; Atp4b; ATPase beta subunit; ATPase Na+/K+ transporting subunit beta 1; ATPase, Na+/K+ transporting, beta 1 polypeptide; Atpb; Atpb-1; ATPBS; Beta 1-subunit of Na(+),K(+)-ATPase; hypothetical protein LOC519758; MGC1798; Na, K-ATPase beta-1 polypeptide; Na,K-ATPase beta 1 subunit; Na+, K+-ATPase; Na+, K+-ATPase beta-subunit; Na+,K+-ATPase beta 1 subunit; Na+/K+ ATPase Beta; NKbeta1; OTTHUMP00000032537; OTTHUMP00000032538; P-type ATPase; Sodium Potassium ATPase; sodium pump subunit beta-1; sodium/potassium ATPase beta subunit; sodium/potassium-dependent ATPase beta-1 subunit; sodium/potassium-dependent ATPase subunit beta-1; sodium/potassium-transporting ATPase beta-1 chain; sodium/potassium-transporting ATPase subunit beta-1; sodium-potassium ATPase subunit beta 1 (non-catalytic); unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11931, 25650, 481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P05026, P07340, P14094 | |
| ATP1B1 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human Sodium Potassium ATPase Beta 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.