Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP1B1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588526 Supplier Thermo Scientific Supplier No. PA588526

Rabbit Polyclonal Antibody

Immunogen sequence: KPTYQDRVAP PGLTQIPQIQ KTEISFRPND PKSYEAYVLN IVRFLEKYKD SAQRDDMIFE DCGDVPSEPK ERGDFNHERG ERKVCRFKLE WLGNCSGLND ETYGYKEGKP CIIIKLNRVL GFKPKPPKNE SLETYPVMKY NPNVLPVQCT GKRDEDKDKV GNVEYFGLGN SPGFPL; Positive Samples: U-251MG, Mouse brain, Mouse liver, Mouse kidney, Rat kidney, Rat brain; Cellular Location: Cell membrane, Single-pass type II membrane protein

The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 1 subunit. Alternatively spliced transcript variants encoding different isoforms have been described, but their biological validity is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 2.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene ATP1B1
Gene Accession No. P05026, P07340, P14094
Gene Alias adenosinetriphosphatase, ATP1B, ATP1B1, Atp4b, ATPase beta subunit, ATPase Na+/K+ transporting subunit beta 1, ATPase, Na+/K+ transporting, beta 1 polypeptide, Atpb, Atpb-1, ATPBS, Beta 1-subunit of Na(+),K(+)-ATPase, hypothetical protein LOC519758, MGC1798, Na, K-ATPase beta-1 polypeptide, Na,K-ATPase beta 1 subunit, Na+, K+-ATPase, Na+, K+-ATPase beta-subunit, Na+,K+-ATPase beta 1 subunit, Na+/K+ ATPase Beta, NKbeta1, OTTHUMP00000032537, OTTHUMP00000032538, P-type ATPase, Sodium Potassium ATPase, sodium pump subunit beta-1, sodium/potassium ATPase beta subunit, sodium/potassium-dependent ATPase beta-1 subunit, sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1, sodium-potassium ATPase subunit beta 1 (non-catalytic), unnamed protein product
Gene Symbols ATP1B1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 65-240 of human ATP1B1 (NP_0016681)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11931, 25650, 481
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.