Learn More
Invitrogen™ ATP1B2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology.
Specifications
Specifications
| Antigen | ATP1B2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ATP1B2 |
| Gene Accession No. | P14415 |
| Gene Alias | adhesion molecule in glia; adhesion molecule on glia; AMOG; ATP1B2; ATPase Na+/K+ transporting subunit beta 2; ATPase, Na+/K+ transporting, beta 2 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 2; ATPB2; Atpb-2; ATPB2S; glial cell adhesion molecule; Na, K-ATPase beta-2 polypeptide; Na,K-ATPase beta 2 subunit; Na+K+ ATPase beta 2 subunit; RATATPB2S; snoRNA MBI-85; Sodium Potassium ATPase; sodium pump subunit beta-2; sodium/potassium-dependent ATPase beta-2 subunit; sodium/potassium-dependent ATPase subunit beta-2; sodium/potassium-transporting ATPase beta-2 chain; sodium/potassium-transporting ATPase subunit beta-2; sodium-potassium ATPase subunit beta 2 (non-catalytic) |
| Gene Symbols | ATP1B2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.