Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552729 Supplier Invitrogen™ Supplier No. PA552729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVSDHTPKYQ DRLATPGLMI RPKTENLDVI VNVSDTESWD QHVQKLNKFL EPYNDSIQAQ KNDVCRPGRY YEQPDNGVLN YPKRACQFNR TQLGNCSGIG DST Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.

This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ ions across the plasma membrane. The exact function of the beta-2 subunit is not known. Mediates cell adhesion of neurons and astrocytes, and promotes neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP1B2
Gene Accession No. P14415
Gene Alias adhesion molecule in glia; adhesion molecule on glia; AMOG; ATP1B2; ATPase Na+/K+ transporting subunit beta 2; ATPase, Na+/K+ transporting, beta 2 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 2; ATPB2; Atpb-2; ATPB2S; glial cell adhesion molecule; Na, K-ATPase beta-2 polypeptide; Na,K-ATPase beta 2 subunit; Na+K+ ATPase beta 2 subunit; RATATPB2S; snoRNA MBI-85; Sodium Potassium ATPase; sodium pump subunit beta-2; sodium/potassium-dependent ATPase beta-2 subunit; sodium/potassium-dependent ATPase subunit beta-2; sodium/potassium-transporting ATPase beta-2 chain; sodium/potassium-transporting ATPase subunit beta-2; sodium-potassium ATPase subunit beta 2 (non-catalytic)
Gene Symbols ATP1B2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP1B2. Recombinant protein control fragment (Product #RP-90193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.