Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121248 Supplier Invitrogen™ Supplier No. PA5121248
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, NCI-H460, SH-SY5Y. The target is usually found in the following locations: Cell membrane, Single-pass type II membrane protein. Immunogen sequence: DHTPKYQDRL ATPGLMIRPK TENLDVIVNV SDTESWDQHV QKLNKFLEPY NDSIQAQKND VCRPGRYYEQ PDNGVLNYPK RACQFNRTQL GNCSGIGDST HYGYSTGQPC VFIKMNRVIN FYAGANQSMN VTCAGKRDED AENLGNFVMF PANGNIDLMY FPYYGKKFHV NYTQPLVAVK FLNVTPNVEV NVECRINAAN IATDDERDKF AGRVAFKLRI NKT.

This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na+ and K+ ions across the plasma membrane. The exact function of the beta-2 subunit is not known. Mediates cell adhesion of neurons and astrocytes, and promotes neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP1B2
Gene Accession No. P13638, P14231, P14415
Gene Alias adhesion molecule in glia; adhesion molecule on glia; AMOG; ATP1B2; ATPase Na+/K+ transporting subunit beta 2; ATPase, Na+/K+ transporting, beta 2 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 2; ATPB2; Atpb-2; ATPB2S; glial cell adhesion molecule; Na, K-ATPase beta-2 polypeptide; Na,K-ATPase beta 2 subunit; Na+K+ ATPase beta 2 subunit; RATATPB2S; snoRNA MBI-85; Sodium Potassium ATPase; sodium pump subunit beta-2; sodium/potassium-dependent ATPase beta-2 subunit; sodium/potassium-dependent ATPase subunit beta-2; sodium/potassium-transporting ATPase beta-2 chain; sodium/potassium-transporting ATPase subunit beta-2; sodium-potassium ATPase subunit beta 2 (non-catalytic)
Gene Symbols ATP1B2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 68-290 of human ATP1B2 (NP_001669.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11932, 24214, 482
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.