Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP1B4 Antibody (108), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290158APC
Description
ATP1B4 Monoclonal antibody specifically detects ATP1B4 in Human samples. It is validated for ELISASpecifications
ATP1B4 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
23439 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
108 | |
ELISA | |
ATPase, (Na+)/K+ transporting, beta 4 polypeptide, ATPase, Na+/K+ transporting, beta 4 polypeptide, K-ATPase beta-m subunit, K-ATPase subunit beta-m, protein ATP1B4, X, X/potassium-transporting ATPase subunit beta-m | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ATP1B4 (mature form, Accession#: NP_036201.1; Tyr 132-Thr 357). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction