Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATP1B4 Monoclonal Antibody (4D5D4)
GREENER_CHOICE

Catalog No. PIMA530073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530073 Supplier Invitrogen™ Supplier No. MA530073
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ATP1B4.

This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na(+) and K(+) ions across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP1B4
Applications ELISA
Classification Monoclonal
Clone 4D5D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ATP1B4
Gene Accession No. Q9UN42
Gene Alias 1110020B02Rik; Ac2-628; Atp1b4; ATPase Na+/K+ transporting family member beta 4; ATPase, (Na+)/K+ transporting, beta 4 polypeptide; ATPase, Na+/K+ transporting, beta 4 polypeptide; BetaM; Na,K-ATPase beta m-subunit; protein ATP1B4; X,K-ATPase beta-m subunit; x,K-ATPase subunit beta-m; X/potassium-transporting ATPase subunit beta-m
Gene Symbols ATP1B4
Host Species Mouse
Immunogen Recombinant Human ATP1B4 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.